Skip to Content
Merck
All Photos(1)

Key Documents

EP5405000042

Eppendorf® Centrifuge 5425

keypad control, with Rotor FA-24x2 including aerosol-tight QuickLock lid, AC/DC input 120 V

Synonym(s):

Centrifuge 5425, centrifuge laboratory, lab centrifuge, micro centrifuge

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161701

AC/DC input

120 V AC, US 3-pin plug

feature

CSA approved
to hold 24 1.5/2.0 mL tubes
with Rotor FA-24x2 including aerosol-tight QuickLock lid
keypad control

manufacturer/tradename

Eppendorf® 5405000042

parameter

15060 rpm speed (Max Speed )

technique(s)

centrifugation: suitable

capacity

10 x 5.0 mL(max rotor capacity)

Looking for similar products? Visit Product Comparison Guide

General description

The centrifuge is equipped with a monitoring glass that provides visual assistance in checking the rotor′s speed or performing a rotor stop using a stroboscope. The control panel features a display and user-friendly keys for seamless operation of the centrifuge.
Centrifuge 5425 is the new laboratory standard with its 24-place capacity and speed up to 21, 330 x G. Focusing on standard applications, it is an essential piece of equipment for modern labs ensuring optimal separation. The centrifuge 5425 serves as a reliable Partner especially for high-throughput users Expecting a top level of ergonomic comfort and provides great flexibility with its 6 rotor options.

Application

Eppendorf® Centrifuge 5425 has been used in the process of DNA preparation for microinjection. It is suitable for the separation of aqueous solutions and suspensions of different densities in approved sample tubes.

Features and Benefits

  • Equipped with a keypad control panel for convenient cleaning
  • Comes with Rotor FA-24x2
  • Non-refrigerated
  • Compatible with 120V power supply and operates on a frequency of 50-60 Hz
  • Reaches a maximum of 21,300 × g or 15,060 rpm
  • Includes 3 program keys for easy selection of user-defined settings
  • Offers over ten different acceleration and braking ramps for precise control

Other Notes

The Centrifuge 5425 is designed for indoor use only. It is essential to adhere to all country-specific safety regulations regarding the operation of electrical equipment in the laboratory.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Generation of Gene Drive Mice for Invasive Pest Population Suppression
Bunting MD, et al.
Methods in Molecular Biology (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service