Skip to Content
Merck
All Photos(1)

Key Documents

NIST1878B

Respirable alpha-quartz

NIST® SRM® 1878b, quantitative X-ray powder diffraction standard

Synonym(s):

Sand, white quartz, Quartz, Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 g

manufacturer/tradename

NIST®

technique(s)

diffraction/scattering: suitable

format

matrix material

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The SRM material was prepared from single crystal nodules of Brazilian quartz that, upon comminution were reduced to a respirable powder with a median particle size of 3.3 µm.

Each unit of SRM 1878b contains approximately 5 g of powder bottled in an argon atmosphere.

SRM 1878B _SDS

SRM 1878B _CoA

Application

Respirable alpha-quartz standard reference material (SRM) is intended for the preparation of calibration standards used in quantitative analyses of α-quartz via X-ray powder diffraction.

Features and Benefits

  • Available with a certificate documenting NIST-certified value of crystalline phase purity and lattice parameter. The information values include particle size distribution, as determined by laser scattering. 
  • Certified values are metrologically traceable to the International System of Units (SI) and SI-derived units of mass fraction.
  • The NIST certificate is provided with storage and use instructions

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L J Williams et al.
Environmental geochemistry and health, 42(7), 2137-2145 (2019-12-18)
Australian Aboriginal populations have unacceptably high rates of bronchiectasis. This disease burden is associated with high rates of detection of pathogenic bacteria, particularly non-typeable Haemophilus influenzae (NTHi). While there is evidence to suggest that exposure to inorganic particulate matter (PM)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service