Key Documents
VPMH203000
Millipore
Viresolve® Pro+ Magnus Holder
Synonym(s):
Viresolve® Pro+ Magnus Holder (for 1 to 3 Devices and 1 to 3 Shields and Shield H)
About This Item
UNSPSC Code:
23151806
eCl@ss:
32031690
material
316L stainless steel component(s) (structural)
Quality Level
manufacturer/tradename
Viresolve®
L × W × H
135 cm (53 in.) × 104 cm (40.9 in.) × 77 cm (30.3 in.)
General description
The Viresolve® Magnus Holders are designed for large volume processing. The Viresolve® Pro+ Magnus Holder is designed to run the Viresolve® Pro Device coupled with either the Viresolve® Pro Shield or Shield H. No product contact eliminates the need for cleaning validations. This cartridge holder, VPMH203000, is capable of accepting 1 to 3 units
Features and Benefits
  • Simple manual hydraulics to clamp units
  • Vertical orientation to minimize footprint and facilitate use (stack, clamp, run)
Legal Information
VIRESOLVE is a registered trademark of Merck KGaA, Darmstadt, Germany