Skip to Content
Merck
All Photos(1)

Key Documents

17-123

Sigma-Aldrich

Anti-Phosphotyrosine Immunoblotting Kit (4G10 , HRP conjugate)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.41

clone

4G10
monoclonal

Quality Level

species reactivity

mouse

technique(s)

western blot: suitable

Components

Anti-Phosphotyrosine, clone 4G10®, HRP conjugate (Cat.# 16-105) Phosphotyrosine control (EGF-stim A431 cell lysate) (Cat.# 12-110) Immunoblot Blocking Reagent (Cat.# 20-200)

Quality

routinely evaluated in immunoblot on EGF-stimulated A431 cell lysates

Legal Information

4G10 is a registered trademark of Upstate Group, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service