Skip to Content
Merck
All Photos(2)

Key Documents

07-2189

Sigma-Aldrich

Anti-CAPZIP Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

RCSD domain containing 1, capZ-interacting protein, Protein kinase substrate CapZIP, protein kinase substrate MK2S4, RCSD domain-containing protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... RCSD1(92241)

General description

CAPZIP (CapZ-interacting protein) is highly expressed in immune cells and muscle and uniquely interacts with CapZ, an F-actin capping protein, in splenocytes. Phosphorylation of CAPZIP is thought to regulate CapZ ability to remodel actin filament assembly. CAPZIP may also contain a CPI (capping protein interaction) motif that seems to restrict capping protein to a low-affinity, filament-binding conformation.

Immunogen

GST-tagged recombinant protein corresponding to human CAPZIP.

Application

Anti-CAPZIP detects levels of CAPZIP proteins & has been published & validated for use in WB & IP.
Immunoprecipitation Analysis: 10 µg from a previous lot immunoprecipitated CAPZIP from Jurkat cell lysate.
Other homologies: Mouse (64% sequence homology). Rat (58% sequence homology).
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling

Quality

Evaluated by Western Blot in Jurkat cell lysate.

Western Blot Analysis: 1:250 dilution of this antibody detected CAPZIP on 10 µg of Jurkat cell lysate.

Target description

~70 kDa observed. The native form of CAPZIP runs anomalously, with a molecular mass of 70 kDa (much slower than its real molecular mass). The 47 kDa band may be non-specific, a splice variant or a degradation product. (Eyers, C.E., et al. (2005). Biochem J. 389(1):127-135.). An uncharacterized band appears at ~18 kDa

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Jurkat cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service