Key Documents
Z742110
Sigma-Aldrich
KitAlysis 24-well reaction block replacement film
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
Quality Level
General description
KitAlysis 24-well Reaction Block replacement films are for use with the KitAlysis 24-well Reaction Block (Z742107) as a part of the KitAlysis High-Throughput Screening Platform. Replacement films are designed for single use and should be replaced between runs for optimal results.
KitAlysis 24-well Reaction Block required for operation (sold separately). Best when used with KitAlysis Benchtop Inertion Box (sold separately).
Legal Information
6538 is a trademark of American Type Culture Collection
KitAlysis is a trademark of Sigma-Aldrich Co. LLC