Key Documents
DWK73790-15
KIMBLE® centrifuge tube
with plain rim finish, conical bottom clear glass, capacity (15 mL), without cap, non-sterile
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
material
V-bottom bottom
conical bottom clear glass
transparent borosilicate glass tube
sterility
non-sterile
feature
without cap
packaging
case of 125 ea shrink wrapped in modular tray
manufacturer/tradename
Kimble® 73790-15
capacity
(15 mL)
fitting
neck (Pushfit)
General description
Plain, disposable centrifuge tube can withstand centrifugation up to 2,980 RCF. Closures are not supplied
  • Conical bottom
  • The 5, 10, 15 sizes have a finished top for a snap-cap closure (not supplied)
  • The 50mL size has a tooled top
  • Shrink wrapped in modular trays to keep them clean and safe in transmit
  • Manufactured from 51 expansion, low extractable borosilicate glass conforming to USP Type 1 and ASTM E438, Type 1, Class B requirements
Screw Thread Disposable Centrifuge Tubes
  • These tubes are suitable for tissue culture work and general bacteriological use.
  • Supplied without caps
  • No marking spot
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences