Merck
  • A genome-wide association study of social and non-social autistic-like traits in the general population using pooled DNA, 500 K SNP microarrays and both community and diagnosed autism replication samples.
A genome-wide association study of social and non-social autistic-like traits in the general population using pooled DNA, 500 K SNP microarrays and both community and diagnosed autism replication samples.
Behavior genetics (2009-12-17)
Angelica Ronald, Lee M Butcher, Sophia Docherty, Oliver S P Davis, Leonard C Schalkwyk, Ian W Craig, Robert Plomin
ABSTRACT
Two separate genome-wide association studies were conducted to identify single nucleotide polymorphisms (SNPs) associated with social and nonsocial autistic-like traits. We predicted that we would find SNPs associated with social and non-social autistic-like traits and that different SNPs would be associated with social and nonsocial. In Stage 1, each study screened for allele frequency differences in approximately 430,000 autosomal SNPs using pooled DNA on microarrays in high-scoring versus low-scoring boys from a general population sample (N = approximately 400/group). In Stage 2, 22 and 20 SNPs in the social and non-social studies, respectively, were tested for QTL association by individually genotyping an independent community sample of 1,400 boys. One SNP (rs11894053) was nominally associated (P < .05, uncorrected for multiple testing) with social autistic-like traits. When the sample was increased by adding females, 2 additional SNPs were nominally significant (P < .05). These 3 SNPs, however, showed no significant association in transmission disequilibrium analyses of diagnosed ASD families.