Merck
  • Expression of UDP-N-acetyl-D-galactosamine: polypeptide N-acetylgalactosaminyltransferase-6 in gastric mucosa, intestinal metaplasia, and gastric carcinoma.
Expression of UDP-N-acetyl-D-galactosamine: polypeptide N-acetylgalactosaminyltransferase-6 in gastric mucosa, intestinal metaplasia, and gastric carcinoma.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (2008-10-16)
Joana Gomes, Nuno T Marcos, Nora Berois, Eduardo Osinaga, Ana Magalhães, João Pinto-de-Sousa, Raquel Almeida, Fátima Gärtner, Celso A Reis
ABSTRACT
Aberrant mucin O-glycosylation is often observed in cancer and is characterized by the expression of immature simple mucin-type carbohydrate antigens. UDP-N-acetyl-d-galactosamine:polypeptide N-acetylgalactosaminyltransferase-6 (ppGalNAc-T6) is one of the enzymes responsible for the initial step in O-glycosylation. This study evaluated the expression of ppGalNAc-T6 in human gastric mucosa, intestinal metaplasia, and gastric carcinomas. Our results showed that ppGalNAc-T6 is expressed in normal gastric mucosa and in intestinal metaplasia. A heterogeneous expression and staining pattern for this enzyme was observed in gastric carcinomas. ppGalNAc-T6 was expressed in 79% of the cases, and its expression level was associated with the presence of venous invasion. Our results provide evidence that ppGalNAc-T6 is an IHC marker associated with venous invasion in gastric carcinoma and may contribute to the understanding of the molecular mechanisms that underlie aberrant glycosylation in gastric carcinogenesis and in gastric carcinoma.