Merck
Design and synthesis of marine fungal phthalide derivatives as PPAR-γ agonists.
Bioorganic & medicinal chemistry (2012-07-24)
Bin Xiao, Jun Yin, Minhi Park, Juan Liu, Jian Lin Li, Eun La Kim, Jongki Hong, Hae Young Chung, Jee H Jung
ABSTRACT
On the basis of a marine fungal phthalide (paecilocin A) skeleton, we synthesized 20 analogs and evaluated them for peroxisome proliferator-activated receptor gamma (PPAR-γ) binding and activation. Among these analogs, 6 and 7 had significant PPAR-γ binding activity, and 7 showed further PPAR-γ activation in rat liver Ac2F cells. In docking simulation, 7 formed H bonds with key amino acid residues of the PPAR-γ binding domain, and the overall positioning was similar to rosiglitazone. This new phthalide derivative is considered an interesting new molecular class of PPAR-γ ligands.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Phthalide, 98%