Documenti
RFA453011-EP
Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges
FA-45-30-11, aerosol tight, to hold, 30 x 1.5/2.0 mL tubes, lid
About This Item
Codice UNSPSC:
41103909
Caratteristiche
lid
to hold 30 x 1.5/2.0 mL tubes
aerosol tight
fixed angle rotor (microtubes)
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Eppendorf® 022637100 (NA)
Eppendorf® 5804726006 (EU)
Parametri
20,800 × g max. RCF
tecniche
centrifugation: suitable
Compatibilità
for use with centrifuge models 5804/5804R/5810/5810R
Note legali
Eppendorf is a registered trademark of Eppendorf AG