Z685526
Vacuubrand chemistry pumping units, manual control, Model PC 101NT
AC/DC input 230 V AC, UK plug
Sinonimo/i:
Vacuubrand 20736403, diaphragm pump
Scegli un formato
3.390,00 €
Scegli un formato
About This Item
3.390,00 €
Prodotti consigliati
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Vacuubrand 733002
Parametri
2.0-2.3 m3/h pumping speed
7 mbar vacuum
Descrizione generale
The manual flow control valve regulates the effective pumping speed at the vacuum connection; the vacuum manometer offers an analog vacuum display. The separator at the inlet (AK), made of glass with a protective coating, retains particles and liquid droplets. The exhaust waste vapor condenser enables near-100-percent solvent recovery.
- Outstanding chemical resistance and superior vapor tolerance
- Exceptionally high performance even at low vacuum
- Excellent ultimate vacuum even with gas ballast
- Manual vacuum control, analog vacuum display
- Excellent environmental friendliness due to efficient solvent recovery
Supplied with MD 4C, manual flow control valve, analogue vacuum gauge, inlet catchpot and exhaust waste vapour condenser certification (NRTL).
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)