Documenti
TEM-G200CU
Sigma-Aldrich
Glider Grids Standard Copper Square Mesh
size 200 mesh, vial of 100
Sinonimo/i:
SQUARE 200 MESH COPPER 100/VIAL
About This Item
Codice UNSPSC:
41111761
NACRES:
MA.24
Confezionamento
vial of 100
Misura
200 mesh
Descrizione generale
Gilder Grids Standard Square Mesh: Copper Grid, 200 mesh, 3.05mm diameter, 0.7 mil (18μm) thickness. Pitch 125μm, Hole 90μm, Bar 35μm