TEM-G1000HHCU
1000 Mesh Hexagonal Copper Grid
vial of 25
Sinonimo/i:
G1000HH-CU HEXAGONAL MESH
Confezionamento
vial of 25
Categorie correlate
Descrizione generale
High Mesh Value Standard Hexagonal Mesh: Copper Grid, 1000 mesh, 3.05mm diameter, 0.7 mil (18μm) thickness. Pitch 25 μm, hole ~19μm, bar ~6μm.