Key Documents
SYNF472950A
Synthware Squibb-style separatory funnel with PTFE stopcock and PTFE stopper
500 mL, top joint: ST/NS 29/42
Sinonimo/i:
Synthware F472950A, glass separatory funnel
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.51
Giunto
joint: ST/NS top29/42
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F472950A
Capacità
500 mL
500 mL
Descrizione generale
Supplied with a Glass or PTFE stopper
Note legali
Synthware is a trademark of Kemtech America Inc.