Documenti
CLS9985125
Pyrex® watch glasses
O.D. 125 mm
Sinonimo/i:
Corning® 9985
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Produttore/marchio commerciale
Corning 9985-125
D.E.
125 mm
Descrizione generale
Heavy-wall, borosilicate glass with fire-polished edge provides durability. May be slightly out-of-round due to O.D. variability.
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.