Documenti
CLS4965250
Pyrex® round-bottom three neck angle type distilling flask
capacity 250 mL
Sinonimo/i:
bottom flask, flask round, flask round bottom
About This Item
Codice UNSPSC:
41104812
NACRES:
NB.43
Materiali
borosilicate glass
round bottom flask
three-neck (Vertical)
Giunto
center joint ST/NS24/40
side joint ST/NS24/40
Sterilità
non-sterile
Caratteristiche
closure type Standard Taper joint
Confezionamento
pack of 1
case of 4
Produttore/marchio commerciale
Corning 4965-250
Capacità
250 mL
Note legali
PYREX is a registered trademark of Corning, Inc.