Merck
HomeMammalian Cell CultureMedium 199 Formulation
Medium 199 Formulation
In 1950, J.F. Morgan conducted experiments with various combinations of vitamins, amino acids, and other factors to develop the novel culture media Medium 199. When properly supplemented, Medium 199 (M199) has broad species applicability, particularly for cultivation of non-transformed cells. It is widely used in virology, vaccine production, and in vitro cultivation of primary explants of mouse pancreatic epithelial and rat lens tissues.
ComponentM4530
[1x] g/L
M5017
g/L
M7528
[1x] g/L
M7653
[1x] g/L
M9163
[10x] g/L
Inorganic Salts
Calcium Chloride0.20.20.20.13961.396
Ferric Nitrate • 9H2O0.000720.000720.000720.000720.0072
Magnesium Sulfate (anhydrous)0.097670.097670.097670.097670.9767
Potassium Chloride0.40.40.40.44
Potassium Phosphate Monobasic0.060.6
Sodium Acetate (anhydrous)0.050.050.050.050.5
Sodium Bicarbonate2.22.20.35
Sodium Chloride6.86.86880
Sodium Phosphate Dibasic (anhydrous)0.047880.4788
Sodium Phosphate Monobasic (anhydrous)0.1220.1220.122
Amino Acids
L-Alanine0.0250.0250.0250.0250.25
L-Arginine • HCl0.070.070.070.070.7
L-Aspartic Acid0.030.030.030.030.3
L-Cysteine • HCl • H2O0.000110.000110.000110.000110.0011
L-Cystine • 2HCl0.0260.0260.0260.0260.26
L-Glutamic Acid0.06680.06680.06680.06680.668
L-Glutamine0.10.1
Glycine0.050.050.050.050.5
L-Histidine • HCl • H2O0.021880.021880.021880.021880.2188
Hydroxy-L-Proline0.010.010.010.010.1
L-Isoleucine0.020.020.020.020.2
L-Leucine0.060.060.060.060.6
L-Lysine • HCl0.070.070.070.070.7
L-Methionine0.0150.0150.0150.0150.15
L-Phenylalanine0.0250.0250.0250.0250.25
L-Proline0.040.040.040.040.4
L-Serine0.0250.0250.0250.0250.25
L-Threonine0.030.030.030.030.3
L-Tryptophan0.010.010.010.010.1
L-Tyrosine • 2Na • 2H2O0.057660.057660.057660.057660.5766
L-Valine0.0250.0250.0250.0250.25
Vitamins
Ascorbic Acid • Na0.00005660.00005660.00005660.00005660.000566
D-Biotin0.000010.000010.000010.000010.0001
Calciferol0.00010.00010.00010.00010.001
Choline Chloride0.00050.00050.00050.00050.005
Folic Acid0.000010.000010.000010.000010.0001
Menadione (sodium bisulfite)0.0000160.0000160.0000160.0000160.00016
myo-Inositol0.000050.000050.000050.000050.0005
Niacinamide0.0000250.0000250.0000250.0000250.00025
Nicotinic Acid0.0000250.0000250.0000250.0000250.00025
p-Aminobenzoic Acid0.000050.000050.000050.000050.0005
D-Pantothenic Acid (hemicalcium)0.000010.000010.000010.000010.0001
Pyridoxal • HCl0.0000250.0000250.0000250.0000250.00025
Pyridoxine • HCl0.0000250.0000250.0000250.0000250.00025
Retinol Acetate0.000140.000140.000140.000140.0014
Riboflavin0.000010.000010.000010.000010.0001
DL-α-Tocopherol Phosphate • Na0.000010.000010.000010.000010.0001
Thiamine • HCl0.000010.000010.000010.000010.0001
Other
Adenine Sulfate0.010.010.010.010.1
Adenosine Triphosphate • 2Na0.0010.0010.0010.0010.01
Adenosine Monophosphate • Na0.00023850.00023850.00023850.00023850.002385
Cholesterol0.00020.00020.00020.00020.002
Deoxyribose0.00050.00050.00050.00050.005
Glucose111110
Glutathione (reduced)0.000050.000050.000050.000050.0005
Guanine • HCl0.00030.00030.00030.00030.003
HEPES5.958
Hypoxanthine0.00030.00030.00030.00030.003
Phenol Red • Na0.02130.02130.02130.02130.0213
Polyoxyethylenesorbitan Monooleate (TWEEN 80)0.020.020.020.020.2
Ribose0.00050.00050.00050.00050.005
Thymine0.00030.00030.00030.00030.003
Uracil0.00030.00030.00030.0003
Xanthine • Na0.0003440.0003440.0003440.000344
Add
L-Glutamine0.110.1 at 1×
Sodium Bicarbonate2.20.35 at 1×
Products
Sorry, an unexpected error has occurred
Network error: Failed to fetch