Merck
HomeServicesSoftware & Digital PlatformsMQuant® StripScan – Don’t Guess; Measure
MQuant® StripScan – Don’t Guess; Measure
The MQuant® StripScan mobile application reads the results of MQuant® test strips in just seconds. Leverage this app for in-process control in food and beverage production, disinfection control or environmental testing, for the following parameters:
  • pH test
  • Ascorbic acid test
  • Ammonium test
  • Nitrate testing
  • Nitrite test
  • Peracetic acid test
  • Peroxide test
  • Sulfate test
  • Sulfite test
  • Total hardness
Our MQuant® test strips provide semi-quantitative detection of parameters. Combined with the app, it provides digital precision when reading your results.
Features:
  • Fast, accurate and reproducible result-readouts - you can trust your digital results
  • Effortless data management, traceability, and sharing documentation and results
    • Universal color reference now provided as added value with all compatible test strips at no extra cost
    • Free MQuant® StripScan app available on Android and Apple app stores – download NOW!
How to measure the parameters using the app?
Conduct Tests Using the App
The logo of MQuant® StripScan App for rapid and accurate reading of various chemical test parameters
Download the FREE MQuant® StripScan App
The MQuant® StripScan App is supported on mobile devices (smart phones and tablets) with iOS 15 and Android OS 12 or higher. Build your profile in the app to have your results documented and your data saved.
To download, search for “MQuant® StripScan” either in the Google Play Store or in the Apple App Store or scan the QR code from the MQuant® Card provided with your MQuant® Test.
Download your free mobile app for test strip readout now
A citrus fruit and a beaker containing a liquid sample from the fruit which is subject to Ascorbic acid testing using MQuant® Ascorbic Acid Test Strips, alongside MQuant® Color Reference Card and packages of the test strips
Start the MQuant® StripScan app on your mobile device and have your MQuant® test strip and MQuant® Card provided in product package ready for testing.
The MQuant® StripScan app allows for easy read-out of 10 chemical parameters, see RELATED PRODUCTS below. 
    Chemist dips MQuant® test strip in a liquid contained in a small beaker to test a chemical parameter
    Dip the test strip into your prepared sample for approx. 2 seconds. Shake off excess liquid from the strip and dry the backside of the strip using a paper towel.
      A user selects the chemical parameter on MQuant® StripScan app, tested using the MQuant® Test Strip, for the result readout by the app installed on the smart phone
      Select the relevant test parameter in the app and follow the wait-time indicated by the countdown clock.
        A user properly placing a dipped MQuant® Test Strip on the color reference card prior to reading by the MQuant® StripScan App on a device
        During the countdown, place your dipped test strip centered in the grey area of the MQuant® Card.
          A user reads the chemical parameter test result by scanning the dipped MQuant® Test Strip using the MQuant® StripScan App on a smart phone
          Before the countdown ends, place the smartphone over the MQuant® Card and align the marks on card and screen.
          The read out is triggered automatically.
            A user glances at the test result of the chemical parameter on MQuant® StripScan App installed on a smart phone
            Get the test result directly from the app screen. All test results are saved to your profile. 
              Image shows the packaging boxes of MQuant® Test Strips, the universal reference card, and two devices displaying the MQuant® StripScan App running
              Log-In to the MQuant® Web Portal at https://www.mquant-stripscan.com/login. Here, you can manage your user profile, view and analyze results, and share test data with team members or directly to a LIMS system.
              Related Technical Resources
              Something went wrong, please try again.
              Related Products
              Sorry, an unexpected error has occurred
              Network error: Failed to fetch