Merck
Primary Cell Culture Techniques
Fluorescence microscopy image showcasing primary cell culture with cells stained to highlight various components: nuclei in blue, actin filaments in green, and mitochondria in red.
An Overview of Primary Cells
Primary cells are mammalian cells that are harvested directly from living human or animal tissues. They differ from stem cells in that non-stem primary cells are typically terminally differentiated and are therefore considered closely representative of the tissues from which they are isolated, particularly where characteristics of cancer biology may make cancer cell lines less predictive models of normal systems or noncancer pathologies. Common primary cell phenotypes include epithelial, endothelial, keratinocytes, melanocytes, fibroblasts, osteoblasts, myocytes, and hepatocytes.   
Something went wrong, please try again.
Featured Categories
A series of five laboratory bottles containing red liquid arranged in a line with a graduated cylinder partially filled with the same red liquid to the right, set against a white background.
Classical Media & Buffers
Choose from a range of optimized cell culture media formulations, including DMEM, RPMI-1640, and serum-free options for diverse cellular needs.
Shop Products
A laboratory scene showing a scientist in a white lab coat standing in the background. In the foreground, various laboratory equipment and containers with pink liquid are arranged on a stainless steel surface. The lab is well-lit, with modern furnishings and equipment visible in the background.
FBS - Fetal Bovine Serum & Fetal Calf Serum
Rich in growth-promoting factors, Fetal bovine serum (FBS) or fetal calf serum (FCS) is an...
Shop Products
Milli-Q® IQ 7003/05/10/15 Ultrapure and Pure Water Purification System - Type 1 and Type 2 lab water purification system with production and tank units plus pure and ultrapure water dispensers.
Milli-Q® Benchtop Lab Water Purification Systems
Milli-Q® systems offer innovative water purification technologies engineered to...
Shop Products
Fluorescent staining of cultured mammalian cells
Mammalian Cell Lines
We offer authenticated, contaminant-free cell lines, many in partnership with ECACC; critical tools for protein, antibody, virus, and vaccine production.
Shop Products
How Primary Cells Differ from Immortalized Cell Lines
Like cell lines derived from tumors or immortalized by other methods, primary cells from human or animal sources provide a method for modeling biological systems. Because they are typically viable for fewer doublings, primary cells are less susceptible to the loss of tissue-specific traits that can characterize cells cultured over long periods of time and repeated passages in vitro. Primary cells may be documented with donor characteristics, including age, gender, and disease state where applicable, and may therefore prove useful models for the development of personalized medicine approaches.
Sourcing and culturing primary cells
Research institutions associated with hospitals may have access to primary tissues and cells. Primary cell suppliers offer cell lots from individual donors or cells pooled from multiple donors. Reputable suppliers or patient collection protocols should include documentation demonstrating ethical oversight for the isolation of cells from human donors, as well as validation of cell type. Primary cells differ from mammalian cell lines in that they are generally more fastidious, requiring extra nutrients, particularly when cultured in media that is low in serum or serum-free. These additional, defined nutrients include cytokines and growth factors.
Document Search
Looking for More Specific Information?
Visit our document search for data sheets, certificates and technical documentation.
Find Documents