HPLC Analysis of Pindolol Enantiomers on Astec® CHIROBIOTIC® T
![HPLC Analysis of Pindolol Enantiomers on Astec® CHIROBIOTIC® T application for HPLC](/deepweb/assets/sigmaaldrich/product/images/382/568/c7fbd9fc-80b7-4223-8ee4-5eb647cc6397/800/c7fbd9fc-80b7-4223-8ee4-5eb647cc6397.jpg)
Materials
analytical column
Product No.
Description
Pricing
Application
Product No.
Description
Pricing
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
15 mM ammonium formate in methanol
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 220 nm
injection
5 μL
sample gas
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC