Merck
GC Analysis of Propylene Oxide Enantiomers on Astec® CHIRALDEX A-TA
GC Analysis of Propylene Oxide Enantiomers on Astec® CHIRALDEX™ A-TA suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX A-TA Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
standard
Product No.
Description
Pricing
(±)-Propylene oxide
puriss. p.a., ≥99.5% (GC)
(±)-Propylene oxide
analytical standard
(S)-(−)-Propylene oxide
99%
(R)-(+)-Propylene oxide
99%
(±)-Propylene oxide
ReagentPlus®, ≥99%
Propylene oxide-d6
≥98 atom % D, 98% (CP), contains hydroquinone as stabilizer
(±)-Propylene oxide
SAJ special grade, ≥99.0%
(±)-Propylene oxide
≥99.5%
CONDITIONS
column
Astec CHIRALDEX A-TA, 30 m x 0.25 mm I.D., 0.12 μm (73003AST)
oven
30 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 24 psi
injection
3 μL, 80:1 split
sample
peak 1: R(+)-propylene oxide
peak 2: S(-)-propylene oxide
Description
Analysis Note
Can also substitute CHIRALDEX B-DP for this separation.
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC