GC Analysis of Alkylamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX™ B-TA
![GC Analysis of Alkylamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX™ B-TA suitable for GC](/deepweb/assets/sigmaaldrich/product/images/323/484/f96fbb30-e89b-4ead-9e73-d6594bfc1a21/800/f96fbb30-e89b-4ead-9e73-d6594bfc1a21.jpg)
CONDITIONS
column
Astec CHIRALDEX B-TA, 30 m x 0.25 mm I.D., 0.12 μm (73023AST)
gradient
80 °C, 2 °C/min to 110 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
injection
0.6 μL, 80:1 split
sample
2.5 mg/mL each racemate in methanol (N-TFA derivatives)
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC