Merck
Adsorption Geometry and Energy Level Alignment at the PTCDA/TiO
The journal of physical chemistry. B (2017-06-21)
Sylvie Rangan, Charles Ruggieri, Robert Bartynski, José Ignacio Martínez, Fernando Flores, José Ortega
ABSTRACT
The adsorption geometry and energy alignment at the PTCDA/TiO
Sigma-Aldrich
PTCDA, 97%
Pricing and availability is not currently available.