Merck
  • The Apoptogenic Toxin AIP56 Is Secreted by the Type II Secretion System of Photobacterium damselae subsp. piscicida.
The Apoptogenic Toxin AIP56 Is Secreted by the Type II Secretion System of Photobacterium damselae subsp. piscicida.
Toxins (2017-11-15)
Ana do Vale, Cassilda Pereira, Carlos R Osorio, Nuno M S dos Santos
ABSTRACT
AIP56 (apoptosis-inducing protein of 56 kDa) is a key virulence factor of
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution