Merck
  • Pilot study of liposome-encapsulated onabotulinumtoxina for patients with overactive bladder: a single-center study.
Pilot study of liposome-encapsulated onabotulinumtoxina for patients with overactive bladder: a single-center study.
European urology (2014-02-22)
Hann-Chorng Kuo, Hsin-Tzu Liu, Yao-Chi Chuang, Lori A Birder, Michael B Chancellor
ABSTRACT
Intradetrusor onabotulinumtoxinA (BoNT-A) injection benefits overactive bladder (OAB) patients, but increased postvoid residual (PVR) urine volume and urinary tract infection (UTI) remain risks. Intravesical instillation of liposomal BoNT-A instead of injection could prevent such adverse events. To evaluate instillation of liquid liposomal BoNT-A (Lipotoxin) for the treatment of OAB and to determine its mechanism of action. A double-blind randomized parallel controlled pilot trial in 24 OAB patients at a single tertiary center. Patients were randomly assigned to intravesical instillation of Lipotoxin containing 80 mg liposomes and 200 U BoNT-A or normal saline (N/S). Patients were retreated with Lipotoxin 1 mo later if they failed the first treatment. Voiding diaries, OAB symptom scores, urodynamic studies, and adverse events were monitored. The primary end point was change of total urinary frequency per 3 d at 1 mo after treatment. Immunohistochemistry and Western blotting for synaptic vesicle glycoprotein 2A (SV2A) and synaptosomal-associated protein, 25 kDa (SNAP25) were performed at baseline and 3 mo after treatment. The Wilcoxon rank sum test and Wilcoxon signed rank test were used for statistical analysis. At 1 mo after treatment, the change of urinary frequency per 3 d significantly improved in the Lipotoxin group (n=12; median: -6.50; interquartile range [IQR]: -18.3 to -0.25; p=0.008) but not in the N/S group. (n=12.0; IQR: -7.75 to 8.0; p=0.792). Urgency episodes also showed a significant decrease in the Lipotoxin group (-12.0; IQR: -20.3 to -2.75; p=0.012) but not in the N/S group (-1.0; IQR: -11.0 to 2.5; p=0.196). SV2A and SNAP25 were expressed in urothelial cells and suburothelial tissues. However, the protein expression did not significantly differ between responders and nonresponders at 3 mo after treatment. Intravesical Lipotoxin instillation effectively reduced frequency episodes 1 mo after treatment in OAB patients without any increase in PVR or risk of UTI. We demonstrated that intravesical Lipotoxin instillation reduced frequency episodes at 1 mo in overactive bladder patients. This procedure is safe, without an increase in postvoid residual or the risk of urinary tract infection.
Sigma-Aldrich
Anti-SV2A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.