Merck
  • Ursolic acid affords antidepressant-like effects in mice through the activation of PKA, PKC, CAMK-II and MEK1/2.
Ursolic acid affords antidepressant-like effects in mice through the activation of PKA, PKC, CAMK-II and MEK1/2.
Pharmacological reports : PR (2017-11-13)
Ana B Ramos-Hryb, Mauricio P Cunha, Francis L Pazini, Vicente Lieberknecht, Rui D S Prediger, Manuella P Kaster, Ana Lúcia S Rodrigues
ABSTRACT
Ursolic acid has been shown to display antidepressant-like effects in mice through the modulation of monoaminergic systems. In this study, we sought to investigate the involvement of signaling pathways on the antidepressant-like effects of ursolic acid. Mice were treated orally with ursolic acid (0.1mg/kg) and, 45min later they received the followings inhibitors by intracerebroventricular route: H-89 (PKA inhibitor, 1μg/mouse), KN-62 (CAMK-II inhibitor, 1μg/mouse), chelerythrine (PKC inhibitor, 1μg/mouse), U0126 (MEK1/2 inhibitor, 5μg/mouse), PD98059 (MEK1/2 inhibitor, 5μg/mouse), wortmannin (PI3K irreversible inhibitor, 0.1μg/mouse) or LY294002 (PI3K inhibitor, 10 nmol/mouse). Immobility time of mice was registered in the tail suspension test (TST). The anti-immobility effect of ursolic acid in the TST was abolished by the treatment of mice with H-89, KN-62, chelerythrine, U0126 or PD98059, but not with wortmannin or LY294002. These results suggest that activation of PKA, PKC, CAMK-II, MEK1/2 may underlie the antidepressant-like effects of ursolic acid.
Sigma-Aldrich
KN-62, ≥95%, powder
Pricing and availability is not currently available.