Merck
  • Establishment of higher passage PER.C6 cells for adenovirus manufacture.
Establishment of higher passage PER.C6 cells for adenovirus manufacture.
Biotechnology progress (2007-12-07)
Marina Berdichevsky, Marie-Pierre Gentile, Benjamin Hughes, Peter Meis, Joseph Peltier, Ilse Blumentals, John Auniņs, Nedim Emil Altaras
ABSTRACT
PER.C6 cells, an industrially relevant cell line for adenovirus manufacture, were extensively passaged in serum-free suspension cell culture to better adapt them to process conditions. The changes in cell physiology that occurred during this passaging were characterized by investigating cell growth, cell size, metabolism, and cultivation of replication-deficient adenovirus. The changes in cell physiology occurred gradually as the population doubling level, the number of times the cell population had doubled, increased. Higher passage PER.C6 (HP PER.C6) proliferated at a specific growth rate of 0.043 h(-1), 2-fold faster than lower passage PER.C6, and were capable of proliferation from lower inoculation cell densities. HP PER.C6 cell volume was 16% greater, and cellular yields on glucose, lactate, oxygen, and amino acids were greater as well. In batch cultures, HP PER.C6 cells volumetrically produced 3-fold more adenovirus, confirmed with three different constructs. The increase in productivity was also seen on a cell-specific basis. Although HP PER.C6 were more sensitive to the "cell density effect", requiring lower infection cell densities for optimal specific productivity, they proliferated more after infection than lower passage PER.C6, increasing the number of cells available for virus production. The extensive passaging established HP PER.C6 cells with several desirable attributes for adenovirus manufacture.
Product Description
SAFC
EX-CELL® GTM-3, Animal-component free, liquid, sterile-filtered, suitable for cell culture
Pricing and availability is not currently available.