Merck
  • Structural and spatial chromatin features at developmental gene loci in human pluripotent stem cells.
Structural and spatial chromatin features at developmental gene loci in human pluripotent stem cells.
Nature communications (2017-11-22)
Hiroki Ikeda, Masamitsu Sone, Shinya Yamanaka, Takuya Yamamoto
ABSTRACT
Higher-order chromatin organization controls transcriptional programs that govern cell properties and functions. In order for pluripotent stem cells (PSCs) to appropriately respond to differentiation signals, developmental gene loci should be structurally and spatially regulated to be readily available for immediate transcription, even though these genes are hardly expressed in PSCs. Here, we show that both chromatin interaction profiles and nuclear positions at developmental gene loci differ between human somatic cells and hPSCs, and that changes in the chromatin interactions are closely related to the nuclear repositioning. Moreover, we also demonstrate that developmental gene loci, which have bivalent histone modifications, tend to colocalize in PSCs. Furthermore, this colocalization requires PRC1, PRC2, and TrxG complexes, which are essential regulatory factors for the maintenance of transcriptionally poised developmental genes. Our results indicate that higher-order chromatin regulation may be an integral part of the differentiation capacity that defines pluripotency.
Sigma-Aldrich
Dextran sulfate sodium salt from Leuconostoc spp., for molecular biology, average Mw >500,000 (dextran starting material), contains 0.5-2% phosphate buffer
Sigma-Aldrich
Anti-WDR5 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-EED Antibody, from rabbit