Merck
  • Maintaining postharvest quality of cold stored 'Hass' avocados by altering the fatty acids content and composition with the use of natural volatile compounds - methyl jasmonate and methyl salicylate.
Maintaining postharvest quality of cold stored 'Hass' avocados by altering the fatty acids content and composition with the use of natural volatile compounds - methyl jasmonate and methyl salicylate.
Journal of the science of food and agriculture (2017-04-28)
Marcin Glowacz, Malick Bill, Peter P Tinyane, Dharini Sivakumar
ABSTRACT
Low temperatures are often used to reduce metabolic processes and extend the storage life of fruit; however, in the case of avocado, a temperature below 3 °C will often result in the development of physiological disorders associated with chilling injury. The objective of this study was to investigate the ability of methyl jasmonate (MeJA) and methyl salicylate (MeSA) vapours to alleviate chilling injury in 'Hass' avocado fruit kept at 2 °C for 21 days followed by 6-7 days of shelf-life at 20 °C, simulating supply chain conditions. The incidence and severity of chilling injury were significantly reduced in MeJA- and MeSA-exposed fruit, especially at 100 µmol L MeJA and MeSA have the potential for being used with 'Hass' avocado fruit shipped at low temperature to reduce its susceptibility to chilling injury. © 2017 Society of Chemical Industry.
Sigma-Aldrich
Tetrahydropyran, anhydrous, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
D-Mannoheptulose, ≥99.0% (HPLC)
Pricing and availability is not currently available.