Merck
  • Cloning and functional expression of alkaline alpha-galactosidase from melon fruit: similarity to plant SIP proteins uncovers a novel family of plant glycosyl hydrolases.
Cloning and functional expression of alkaline alpha-galactosidase from melon fruit: similarity to plant SIP proteins uncovers a novel family of plant glycosyl hydrolases.
The Plant journal : for cell and molecular biology (2003-08-29)
Nir Carmi, Genfa Zhang, Marina Petreikov, Zhifang Gao, Yoram Eyal, David Granot, Arthur A Schaffer
ABSTRACT
Raffinose and stachyose are ubiquitous galactosyl-sucrose oligosaccharides in the plant kingdom which play major roles, second only to sucrose, in photoassimilate translocation and seed carbohydrate storage. These sugars are initially metabolised by alpha-galactosidases (alpha-gal). We report the cloning and functional expression of the first genes, CmAGA1 and CmAGA2, encoding for plant alpha-gals with alkaline pH optima from melon fruit (Cucumis melo L.), a raffinose and stachyose translocating species. The alkaline alpha-gal genes show very high sequence homology with a family of undefined 'seed imbibition proteins' (SIPs) which are present in a wide range of plant families. In order to confirm the function of SIP proteins, a representative SIP gene, from tomato, was expressed and shown to have alkaline alpha-gal activity. Phylogenetic analysis based on amino acid sequences shows that the family of alkaline alpha-gals shares little homology with the known prokaryotic and eukaryotic alpha-gals of glycosyl hydrolase families 27 and 36, with the exception of two cross-family conserved sequences containing aspartates which probably function in the catalytic step. This previously uncharacterised, plant-specific alpha-gal family of glycosyl hydrolases, with optimal activity at neutral-alkaline pH likely functions in key processes of galactosyl-oligosaccharide metabolism, such as during seed germination and translocation of RFO photosynthate.