- Early-life immune activation increases song complexity and alters phenotypic associations between sexual ornaments.
Early-life immune activation increases song complexity and alters phenotypic associations between sexual ornaments.
Functional ecology (2018-02-06)
Loren Merrill, Madeleine F Naylor, Merria Dalimonte, Sean McLaughlin, Tara E Stewart, Jennifer L Grindstaff
PMID29398763
ABSTRACT
Early-life adversity can have long-lasting effects on physiological, behavioural, cognitive, and somatic processes. Consequently, these effects may alter an organism's life-history strategy and reproductive tactics.In response to early-life immune activation, we quantified levels of the acute phase protein haptoglobin (Hp) during development in male zebra finches (