Merck
  • A comparison of steroid profiles in the testis and seminal vesicle of the catfish (Heteropneustes fossilis).
A comparison of steroid profiles in the testis and seminal vesicle of the catfish (Heteropneustes fossilis).
Theriogenology (2017-09-25)
Radha Chaube, Surabhi Mishra, Rahul Kumar Singh
ABSTRACT
In the present study, distribution of steroid hormones (estradiol-17β (E
Sigma-Aldrich
21-Deoxycortisol
Pricing and availability is not currently available.