Merck
  • ARHI is a novel epigenetic silenced tumor suppressor in sporadic pheochromocytoma.
ARHI is a novel epigenetic silenced tumor suppressor in sporadic pheochromocytoma.
Oncotarget (2017-11-22)
Dong Wang, Li Song, Liang Wang, Lianmei Zhao, Bai Xiang, Ying Li, Baoen Shan, Jing Liu
ABSTRACT
Pheochromocytoma (PCC) is related to germline mutations in 12 susceptibility genes. Although comparative genomic hybridization array has revealed some putative tumor suppressor genes on the short arm of chromosome 1 that are likely to be involved in PCC tumorigenesis, the molecules involved, except for those encoded by known susceptibility genes, have not been found in the generation of sporadic tumors. In the present work, we first identified that the unmethylated allele of Aplasia Ras homolog member I (
Sigma-Aldrich
5-Aza-2′-deoxycytidine, ≥97%
Sigma-Aldrich
MISSION® esiRNA, targeting human DIRAS3