Merck
  • UV-Induced Hydrogen-Atom-Transfer Processes in 3-Thio-1,2,4-triazole Isolated in Ar and H
UV-Induced Hydrogen-Atom-Transfer Processes in 3-Thio-1,2,4-triazole Isolated in Ar and H
The journal of physical chemistry. A (2017-08-30)
Hanna Rostkowska, Leszek Lapinski, Maciej J Nowak
ABSTRACT
The UV-induced thione → thiol phototautomeric reaction has been studied for monomeric 3-thio-1,2,4-triazole (3-ST) isolated in low-temperature Ar and n-H
Sigma-Aldrich
2-Amino-5-methylpyridine, 99%
Sigma-Aldrich
1H-1,2,3-Triazole, 97%
Sigma-Aldrich
2-(Methylamino)pyridine, 98%