Merck
  • Combining Fungicides and Prospective NPR1-Based "Just-in-Time" Immunomodulating Chemistries for Crop Protection.
Combining Fungicides and Prospective NPR1-Based "Just-in-Time" Immunomodulating Chemistries for Crop Protection.
Frontiers in plant science (2017-10-20)
Xiahezi Kuai, Charles Barraco, Charles Després
ABSTRACT
Each year, crop yield is lost to weeds competing for resources, insect herbivory and diseases caused by pathogens. To thwart these insults and preserve yield security and a high quality of traits, conventional agriculture makes use of improved cultivars combined with fertilizer and agrochemical applications. However, given that regulatory bodies and consumers are demanding environmentally safer agrochemicals, while at the same time resistance to agrochemicals is mounting, it is crucial to adopt a "holistic" approach to agriculture by not excluding any number of management tools at our disposal. One such tool includes chemicals that stimulate plant immunity. The development of this particular type of alternative crop protection strategy has been of great interest to us. We have approached this paradigm by studying plant immunity, specifically systemic acquired resistance (SAR). The deployment of SAR immunity requires the production by the crop plant of an endogenous small molecule metabolite called salicylic acid (SA). Furthermore, immunity can only be deployed if SA can bind to its receptor and activate the genes responsible for the SAR program. The key receptor for SAR is a transcription coactivator called NPR1. Since discovering this NPR1-SA receptor-ligand pair, we have embarked on a journey to develop novel chemistries capable of deploying SAR in the field. The journey begins with the development of a scalable assay to identify these novel chemistries. One such assay, presented here, is based on differential scanning fluorimetry technology and demonstrates that NPR1 is destabilized by binding to SA.
Product Description
Supelco
SYPRO® Orange Protein Gel Stain
Pricing and availability is not currently available.