- Alumina/activated carbon nano-composites: Synthesis and application in sulphide ion removal from water.
Alumina/activated carbon nano-composites: Synthesis and application in sulphide ion removal from water.
Journal of hazardous materials (2017-07-18)
Ushadevi Balasubramani, Ranjithkumar Venkatesh, Sangeetha Subramaniam, Gayathri Gopalakrishnan, Vairam Sundararajan
PMID28715747
ABSTRACT
An investigation of adsorption of sulphide ion (S