Merck
  • Over-expression of BAG-1 in head and neck squamous cell carcinomas (HNSCC) is associated with cisplatin-resistance.
Over-expression of BAG-1 in head and neck squamous cell carcinomas (HNSCC) is associated with cisplatin-resistance.
Journal of translational medicine (2017-09-08)
Shutong Liu, Bo Ren, Hang Gao, Suchan Liao, Ying-Xian Zhai, Shirong Li, Xue-Jin Su, Ping Jin, David Stroncek, Zhixiang Xu, Qinghua Zeng, Yulin Li
ABSTRACT
In order to improve therapy for head and neck squamous cell carcinoma (HNSCC), biomarkers associated with local and/or distant tumor relapses and cancer drug resistance are urgently needed. This study identified a potential biomarker, Bcl-2 associated athanogene-1 (BAG-1), that is implicated in HNSCC insensitive to cisplatin and tumor progression. Primary and advanced (relapsed from parental) University of Michigan squamous cell carcinoma cell lines were tested for sensitivity to cisplatin and gene expression profiles were compared between primary (cisplatin sensitive) and the relapsed (cisplatin resistant) cell lines by using Agilent microarrays. Additionally, differentially expressed genes phosphorylated AKT, and BAG-1, and BCL-xL were evaluated for expression using HNSCC tissue arrays. Advanced HNSCC cells revealed resistant to cisplatin accompanied by increased expression of BAG-1 protein. siRNA knockdown of BAG-1 expression resulted in significant improvement of HNSCC sensitivity to cisplatin. BAG-1 expression enhanced stability of BCL-xL and conferred cisplatin resistant to the HNSCC cells. In addition, high levels of expression of phosphorylated AKT, BAG-1, and BCL-xL were observed in advanced HNSCC compared to in that of primary HNSCC. Increased expression of BAG-1 was associated with cisplatin resistance and tumor progression in HNSCC patients and warrants further validation in larger independent studies. Over expression of BAG-1 may be a biomarker for cisplatin resistance in patients with primary or recurrent HNSCCs and targeting BAG-1 could be helpful in overcoming cisplatin resistance.
Sigma-Aldrich
MISSION® esiRNA, targeting human BAG1
Sigma-Aldrich
S3I-201, ≥97% (HPLC)