Merck
The EspF N-Terminal of Enterohemorrhagic
Frontiers in cellular and infection microbiology (2017-10-07)
Xiangyu Wang, Yanli Du, Ying Hua, Muqing Fu, Cong Niu, Bao Zhang, Wei Zhao, Qiwei Zhang, Chengsong Wan
ABSTRACT
Enterohemorrhagic
Sigma-Aldrich
N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide, ≥97% (HPLC), powder