Merck
  • Bifunctional Boron Phosphate as an Efficient Catalyst for Epoxide Activation to Synthesize Cyclic Carbonates with CO
Bifunctional Boron Phosphate as an Efficient Catalyst for Epoxide Activation to Synthesize Cyclic Carbonates with CO
Chemistry, an Asian journal (2017-06-20)
Zhimin Xue, Xinhui Zhao, Jinfang Wang, Tiancheng Mu
ABSTRACT
Development of inexpensive, easily prepared, non-toxic, and efficient catalysts for the cycloaddition of CO
Sigma-Aldrich
1,2-Epoxyhexane, 97%
Sigma-Aldrich
Cyclohexene oxide, 98%