Merck
  • Nanostructured polysaccharidic microcapsules for intracellular release of cisplatin.
Nanostructured polysaccharidic microcapsules for intracellular release of cisplatin.
International journal of biological macromolecules (2017-02-25)
Viviana Vergaro, Paride Papadia, Paola Petrini, Francesco Paolo Fanizzi, Sandra A De Pascali, Francesca Baldassarre, Laura Pastorino, Giuseppe Ciccarella
ABSTRACT
Carbohydrate polimeric microcapsules were assembled using a LbL approach onto a CaCO
Sigma-Aldrich
Chitosan, medium molecular weight
Pricing and availability is not currently available.