Merck
  • Demonstration of thermo-sensitive tetra-gel with implication for facile and versatile platform for a new class of smart gels.
Demonstration of thermo-sensitive tetra-gel with implication for facile and versatile platform for a new class of smart gels.
Journal of biomaterials science. Polymer edition (2017-04-11)
Akira Matsumoto, Shohei Sato, Tomoko Sakamaki, Mai Sanjo, Miyuki Tabata, Tatsuro Goda, Taka-Aki Asoh, Akihiko Kikuchi, Yuji Miyahara
ABSTRACT
A tertiary branched poly(N-isopropylacrylamide) with controlled molecular weight, distribution and the end amino-functionalization (tetra-PNIPAAm-NH
Sigma-Aldrich
N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine, 99%
Sigma-Aldrich
2-Chloropropionyl chloride, 97%
Sigma-Aldrich
Pentaerythritol tetrakis(2-bromoisobutyrate), 97%
Sigma-Aldrich
Tris[2-(dimethylamino)ethyl]amine, 97%