Merck
  • Functional expression of calcium-permeable canonical transient receptor potential 4-containing channels promotes migration of medulloblastoma cells.
Functional expression of calcium-permeable canonical transient receptor potential 4-containing channels promotes migration of medulloblastoma cells.
The Journal of physiology (2017-06-20)
Wei-Chun Wei, Wan-Chen Huang, Yu-Ping Lin, Esther B E Becker, Olaf Ansorge, Veit Flockerzi, Daniele Conti, Giovanna Cenacchi, Maike D Glitsch
ABSTRACT
The proton sensing ovarian cancer G protein coupled receptor 1 (OGR1, aka GPR68) promotes expression of the canonical transient receptor potential channel subunit TRPC4 in normal and transformed cerebellar granule precursor (DAOY) cells. OGR1 and TRPC4 are prominently expressed in healthy cerebellar tissue throughout postnatal development and in primary cerebellar medulloblastoma tissues. Activation of TRPC4-containing channels in DAOY cells, but not non-transformed granule precursor cells, results in prominent increases in [Ca Aberrant intracellular Ca
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human TRPC4
Sigma-Aldrich
ML204, ≥98% (HPLC)