Merck
  • Anti-Cancer Drug HMBA Acts as an Adjuvant during Intracellular Bacterial Infections by Inducing Type I IFN through STING.
Anti-Cancer Drug HMBA Acts as an Adjuvant during Intracellular Bacterial Infections by Inducing Type I IFN through STING.
Journal of immunology (Baltimore, Md. : 1950) (2017-08-23)
Akshamal Mihiranga Gamage, Kok-Onn Lee, Yunn-Hwen Gan
ABSTRACT
The anti-proliferative agent hexamethylene bisacetamide (HMBA) belongs to a class of hybrid bipolar compounds developed more than 30 y ago for their ability to induce terminal differentiation of transformed cells. Recently, HMBA has also been shown to trigger HIV transcription from latently infected cells, via a CDK9/HMBA inducible protein-1 dependent process. However, the effect of HMBA on the immune response has not been explored. We observed that pretreatment of human peripheral blood mononuclear cells with HMBA led to a markedly increased production of IL-12 and IFN-γ, but not of TNF-α, IL-6, and IL-8 upon subsequent infection with Burkholderia pseudomallei and Salmonella enterica HMBA treatment was also associated with better intracellular bacterial control. HMBA significantly improved IL-12p70 production from CD14+ monocytes during infection partly via the induction of type I IFN in these cells, which primed an increased transcription of the p35 subunit of IL-12p70 during infection. HMBA also increased early type I IFN transcription in human monocytic and epithelial cell lines, but this was surprisingly independent of its previously reported effects on positive transcription elongation factor b and HMBA inducible protein-1. Instead, the effect of HMBA was downstream of a calcium influx, and required the pattern recognition receptor and adaptor STING but not cGAS. Our work therefore links the STING-IRF3 axis to enhanced IL-12 production and intracellular bacterial control in primary monocytes. This raises the possibility that HMBA or related small molecules may be explored as therapeutic adjuvants to improve disease outcomes during intracellular bacterial infections.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
MISSION® esiRNA, targeting human DDX58
Sigma-Aldrich
Resiquimod, ≥98% (HPLC)
Sigma-Aldrich
SAHA, ≥98% (HPLC)