Merck
  • Hepatic estrogen receptor α is critical for regulation of gluconeogenesis and lipid metabolism in males.
Hepatic estrogen receptor α is critical for regulation of gluconeogenesis and lipid metabolism in males.
Scientific reports (2017-05-12)
Shuiqing Qiu, Juliana Torrens Vazquez, Erin Boulger, Haiyun Liu, Ping Xue, Mehboob Ali Hussain, Andrew Wolfe
ABSTRACT
Impaired estrogens action is associated with features of the metabolic syndrome in animal models and humans. We sought to determine whether disruption of hepatic estrogens action in adult male mice could recapitulate aspects of the metabolic syndrome to understand the mechanistic basis for the phenotype. We found 17β-estradiol (E
Sigma-Aldrich
N-Ethyl-N-(2-hydroxy-3-sulfopropyl)-3,5-dimethoxyaniline sodium salt