Merck
Palmdelphin promotes myoblast differentiation and muscle regeneration.
Scientific reports (2017-02-06)
Yaping Nie, Hu Chen, Cilin Guo, Zhuning Yuan, Xingyu Zhou, Ying Zhang, Xumeng Zhang, Delin Mo, Yaosheng Chen
ABSTRACT
Differentiation of myoblasts is essential in the development and regeneration of skeletal muscles to form multinucleated, contractile muscle fibers. However, the process of myoblast differentiation in mammals is complicated and requires to be further investigated. In this study, we found Palmdelphin (Palmd), a cytosolic protein, promotes myoblast differentiation. Palmd is predominantly expressed in the cytosol of myoblasts and is gradually up-regulated after differentiation. Knockdown of Palmd by small interfering RNA (siRNA) in C2C12 markedly inhibits myogenic differentiation, suggesting a specific role of Palmd in the morphological changes of myoblast differentiation program. Overexpression of Palmd in C2C12 enhances myogenic differentiation. Remarkably, inhibition of Palmd results in impaired myotube formation during muscle regeneration after injury. These findings reveal a new cytosolic protein that promotes mammalian myoblast differentiation and provide new insights into the molecular regulation of muscle formation.
Sigma-Aldrich
MISSION® esiRNA, targeting human PALMD
Sigma-Aldrich
Anti-Myogenin Antibody, Chemicon®, from mouse