Merck
  • Cucurbitacin IIa exerts antidepressant-like effects on mice exposed to chronic unpredictable mild stress.
Cucurbitacin IIa exerts antidepressant-like effects on mice exposed to chronic unpredictable mild stress.
Neuroreport (2017-02-28)
Shi-Meng Zhou, Shao-Yu Guan, Le Yang, Liu-Kun Yang, Lu Wang, Hui-Fang Nie, Xiang Li, Ming-Gao Zhao, Qi Yang, Hong Wu
ABSTRACT
Cucurbitacin IIa (CuIIa) is the major active component of the Helmseya amabilis root and is known to have antiviral and anti-inflammatory effects. In this study, we examined the antidepressant-like effects of CuIIa in a mouse model of chronic unpredictable mild stress (CUMS) and investigated the possible underlying mechanisms. To evaluate the antidepressant-like effects of CuIIa on depression-like behaviors, mice were subjected to the open-field test, the elevated plus-maze test, the forced-swimming test, and the tail-suspension test. We found that CuIIa treatment reversed the CUMS-induced behavioral abnormalities. Western blot analyses showed that CUMS significantly decreased brain-derived neurotrophic factor (BDNF) levels, cAMP-response element binding protein (CREB), and calcium/calmodulin-dependent protein kinase II (CaMKII) phosphorylation, and N-methyl-D-aspartate receptor subtype GluN2B and α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid receptor GluA1 expression in the amygdala; in addition, the expression of gamma-aminobutyric acid receptor A subunit α2 was upregulated in CUMS mice. These CUMS-induced changes were all normalized by CuIIa treatment and administration of the BDNF antagonist ANA-12 can block the antidepressant effect of CuIIa. Our findings suggest that the antidepressant-like effects of CuIIa may be exerted by regulation of the CaMKIIα-CREB-BDNF pathway and the balance between excitatory and inhibitory synaptic transmission in the amygdala.
Sigma-Aldrich
ANA-12, ≥98% (HPLC)