Merck
  • Charge tunable thin-film composite membranes by gamma-ray triggered surface polymerization.
Charge tunable thin-film composite membranes by gamma-ray triggered surface polymerization.
Scientific reports (2017-07-02)
Rackel Reis, Mikel C Duke, Blaise L Tardy, Daniel Oldfield, Raymond R Dagastine, John D Orbell, Ludovic F Dumée
ABSTRACT
Thin-film composite poly(amide) (PA) membranes have greatly diversified water supplies and food products. However, users would benefit from a control of the electrostatic interactions between the liquid and the net surface charge interface in order to benefit wider application. The ionic selectivity of the 100 nm PA semi-permeable layer is significantly affected by the pH of the solution. In this work, for the first time, a convenient route is presented to configure the surface charge of PA membranes by gamma ray induced surface grafting. This rapid and up-scalable method offers a versatile route for surface grafting by adjusting the irradiation total dose and the monomer concentration. Specifically, thin coatings obtained at low irradiation doses between 1 and 10 kGy and at low monomer concentration of 1 v/v% in methanol/water (1:1) solutions, dramatically altered the net surface charge of the pristine membranes from -25 mV to +45 mV, whilst the isoelectric point of the materials shifted from pH 3 to pH 7. This modification resulted in an improved water flux by over 55%, from 45.9 to up 70 L.m
Sigma-Aldrich
1-Vinylimidazole, ≥99%
Pricing and availability is not currently available.