Merck
  • Formation, characterization, and mineralization of bound residues of tetrabromobisphenol A (TBBPA) in silty clay soil under oxic conditions.
Formation, characterization, and mineralization of bound residues of tetrabromobisphenol A (TBBPA) in silty clay soil under oxic conditions.
The Science of the total environment (2017-05-10)
Songfeng Wang, Feifei Sun, Yongfeng Wang, Lianhong Wang, Yini Ma, Boris Alexander Kolvenbach, Philippe François-Xavier Corvini, Rong Ji
ABSTRACT
The nature and stability of bound residues (BRs) derived from the widely used brominated flame retardant tetrabromobisphenol A (TBBPA) in fine-textured soil is unknown. We incubated
Sigma-Aldrich
Murashige and Skoog Basal Salt Mixture (MS), powder, suitable for plant cell culture
Pricing and availability is not currently available.