Merck
  • Purinergic P2Y1 Receptors Control Rapid Expression of Plasma Membrane Processes in Hippocampal Astrocytes.
Purinergic P2Y1 Receptors Control Rapid Expression of Plasma Membrane Processes in Hippocampal Astrocytes.
Molecular neurobiology (2016-06-20)
Mariangela Chisari, Angela Scuderi, Lucia Ciranna, Guido Li Volsi, Flora Licata, Maria Angela Sortino
ABSTRACT
Astrocytes regulate neuronal activity and blood brain barrier through tiny plasma membrane branches or astrocytic processes (APs) making contact with synapses and brain vessels. Several transmitters released by astrocytes and exerting their action on several receptor classes expressed by astrocytes themselves influence their physiology. Here we found that APs are dynamically modulated by purines. In live imaging experiments carried out in rat hippocampal astrocytes, Gq-coupled P2Y1 receptor blockade with the selective antagonist MRS2179 (1 μM) or inhibition of its effector phospholipase C using U73122 (3 μM) produced APs retraction, while stimulation of the same receptor with the selective agonist 2MeSADP (100 μM) increased their number. Since astrocytes, among other transmitters, release ATP by several mechanisms including connexin hemichannels, we used the connexin hemichannel inhibitor carbenoxolone (100 μM) and APs retraction was observed. In our system we then measured expression or function of channels important for modulation of volume transmission and K+ buffering, aquaporin-4, and K+ inward rectifying (Kir) channels, respectively. Aquaporin-4 expression level did not change whereas, in whole-cell patch-clamp recordings performed to measure Kir current, we observed an increase in K+ current in all conditions where APs number was reduced. These data are supporting the idea of a dynamic modulation of astrocytic processes by purinergic signal, strengthening the role of purines in brain homeostasis.
Sigma-Aldrich
N6-Methyl-2′-deoxyadenosine
Pricing and availability is not currently available.